siRNA Details
Gene Symbol: LINC00917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024406.1 | 4 | 1303 | |||
GenBank mRNA | AK093244.1 | - | 4 | 1303 | ||
BC043250.1 | - | 3 | 1360 | |||
BC144028.1 | - | 2 | 523 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL137562.1 | - | 1 | 16 | Chr. 16 - 86331844 - 86337556 [-] on Build GRCh38 |
