siRNA Details
Gene Symbol: LINC00917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024406.1 | 7 | 2426 | |||
GenBank mRNA | AK093244.1 | - | 7 | 2426 | ||
BC043250.1 | - | 6 | 2483 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL137562.1 | - | 5 | 1334 | Chr. 16 - 86331844 - 86337556 [-] on Build GRCh38 |
