siRNA Details
Gene Symbol: LINC01366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026945.2 | 2 | 242 | |||
NR_108024.1 | 2 | 223 | ||||
GenBank mRNA | AK093725.1 | - | 2 | 938 | ||
BQ053612.1 | - | 2 | 204 | |||
CD518314.1 | - | 2 | 187 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX649024.1 | - | 1 | 58 | Chr. 2 - 113762423 - 113766911 [+] on Build GRCh38 |
