siRNA Details
Gene Symbol: JHDM1D-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024451.1 | 1 | 770 | |||
GenBank mRNA | AK026372.1 | - | 1 | 672 | ||
DA816865.1 | - | 1 | 327 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: JHDM1D-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024451.1 | 1 | 770 | |||
GenBank mRNA | AK026372.1 | - | 1 | 672 | ||
DA816865.1 | - | 1 | 327 |