siRNA Details
Gene Symbol: LOC100240735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026658.1 | 3 | 708 |
Gene Symbol: LOC107985433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028192.1 | XP_016883681.1 | 3 | 1345 |
