siRNA Details
Gene Symbol: RBMY2EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001574.2 | 5 | 516 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U94388.3 | - | 5 | 516 | Chr. Y - 21395148 - 21401562 [-] on Build GRCh38 |
