siRNA Details
Gene Symbol: LOC100133669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026913.1 | 2 | 552 | |||
GenBank mRNA | BM557790.1 | - | 2 | 552 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI824732.1 | - | 2 | 553 | Chr. 8 - 142981738 - 143018386 [-] on Build GRCh38 |
