siRNA Details
Gene Symbol: LOC100133669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026913.1 | 2 | 468 | |||
GenBank mRNA | BM557790.1 | - | 2 | 468 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100133669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026913.1 | 2 | 468 | |||
GenBank mRNA | BM557790.1 | - | 2 | 468 |