siRNA Details
Gene Symbol: LOC100133669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026913.1 | 1 | 202 | |||
GenBank mRNA | BC007589.1 | - | 1 | 51 | ||
BM557790.1 | - | 1 | 202 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100133669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026913.1 | 1 | 202 | |||
GenBank mRNA | BC007589.1 | - | 1 | 51 | ||
BM557790.1 | - | 1 | 202 |