siRNA Details
Gene Symbol: PLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243079.1 | NP_001230008.1 | 9, 10 | 1128 | ||
GenBank mRNA | AK054808.1 | - | 9, 10 | 1128 | ||
AK122648.1 | - | 8, 9 | 1300 | |||
BC100791.2 | - | 6, 7 | 692 | |||
BC100792.1 | - | 6, 7 | 692 | |||
DQ424898.1 | - | 3, 4 | 412 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI827807.1 | - | 3, 4 | 343 | Chr. 19 - 1528003 - 1535429 [+] on Build GRCh38 |
