siRNA Details
Gene Symbol: LINC01140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026985.1 | 3 | 575 | |||
NR_026986.1 | 2 | 375 | ||||
NR_026987.1 | 2 | 368 | ||||
NR_026988.1 | 2 | 271 | ||||
NR_026989.1 | 2 | 271 | ||||
GenBank mRNA | AL833352.1 | - | 3 | 599 | ||
BC030523.1 | - | 2 | 166 | |||
BC041867.1 | - | 3 | 546 | |||
BI818944.1 | - | 2 | 182 | |||
BM722117.1 | - | 2 | 181 | |||
DA398164.1 | - | 2 | 362 | |||
DA981741.1 | - | 2 | 274 | |||
DB100426.1 | - | 2 | 337 |
