siRNA Details
Gene Symbol: LINC01139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015407.1 | 2 | 488 | |||
GenBank mRNA | BC044843.1 | - | 2 | 488 | ||
BC064529.1 | - | 2 | 494 | |||
BC073894.1 | - | 2 | 465 | |||
BC113688.1 | - | 1 | 204 | |||
BC113690.1 | - | 1 | 204 |
