siRNA Details
Gene Symbol: LOC151174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026925.1 | 3 | 1943 | |||
NR_026926.1 | 3 | 830 | ||||
GenBank mRNA | AF451986.1 | - | 2 | 421 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR738501.1 | - | 1 | 600 | Chr. 2 - 238225013 - 238225728 [-] on Build GRCh38 |
