siRNA Details
Gene Symbol: LOC151174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026925.1 | 2 | 395 | |||
NR_026926.1 | 2 | 395 | ||||
GenBank mRNA | AK057399.1 | - | 2 | 331 | ||
DA518680.1 | - | 2 | 395 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC151174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026925.1 | 2 | 395 | |||
NR_026926.1 | 2 | 395 | ||||
GenBank mRNA | AK057399.1 | - | 2 | 331 | ||
DA518680.1 | - | 2 | 395 |