siRNA Details
Gene Symbol: LINC00207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028409.1 | 5 | 520 | |||
NR_028410.1 | 4 | 489 | ||||
NR_028411.1 | 4 | 453 | ||||
GenBank mRNA | BC144506.1 | - | 4 | 477 | ||
BC144508.1 | - | 5 | 508 | |||
BU585030.1 | - | 4 | 472 |
