siRNA Details
Gene Symbol: LINC00298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015405.1 | 2 | 174 | |||
GenBank mRNA | BC043361.1 | - | 3 | 245 | ||
BC103700.1 | - | 2 | 174 | |||
BC104747.1 | - | 2 | 174 | |||
BC105688.1 | - | 2 | 174 | |||
BC107115.1 | - | 2 | 174 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF338305.1 | - | 3 | 244 | Chr. 2 - 7923917 - 7979309 [-] on Build GRCh38 |
