siRNA Details
Gene Symbol: SNORA16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003035.1 | 11 | ||||
GenBank mRNA | AJ609433.1 | - | 11 |
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092096.1 | - | 1 | 833 | ||
BC034611.1 | - | 2 | 262 |
