siRNA Details
Gene Symbol: LINC00442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026852.1 | 4 | 1374 | |||
GenBank mRNA | AK301801.1 | - | 4 | 1371 | ||
BC047425.1 | - | 4 | 1374 | |||
BC067875.1 | - | 4 | 1374 | |||
BC094789.1 | - | 4 | 1374 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY013853.1 | - | 3 | 454 | Chr. 13 - 19009653 - 19011951 [+] on Build GRCh38 |
