siRNA Details
Gene Symbol: FLVCR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027285.1 | 1 | 231 | |||
NR_027286.1 | 1 | 231 | ||||
GenBank mRNA | AK092887.1 | - | 1 | 181 | ||
AY030238.1 | - | 1 | 170 | |||
AY030239.1 | - | 1 | 170 |
