siRNA Details
Gene Symbol: LOC100134868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004846.3 | 3 | 255 | |||
GenBank mRNA | BC030737.1 | - | 3 | 220 | ||
BC062357.1 | - | 3 | 220 | |||
CR987009.1 | - | 2 | 174 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100134868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004846.3 | 3 | 255 | |||
GenBank mRNA | BC030737.1 | - | 3 | 220 | ||
BC062357.1 | - | 3 | 220 | |||
CR987009.1 | - | 2 | 174 |