siRNA Details
Gene Symbol: LOC100134868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004846.3 | 3, 4 | 304 | |||
GenBank mRNA | BC030737.1 | - | 3, 4 | 269 | ||
BC062357.1 | - | 3, 4 | 269 | |||
CR987009.1 | - | 2, 3 | 223 |
Gene Symbol: ZNF850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193552.1 | NP_001180481.1 | 3, 4 | 303 | ||
GenBank mRNA | BC052603.1 | - | 3, 4 | 303 | ||
BC136266.1 | - | 3, 4 | 241 |
