siRNA Details
Gene Symbol: LOC100134868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004846.3 | 5 | 1441 | |||
GenBank mRNA | BC030737.1 | - | 5 | 1406 | ||
BC062357.1 | - | 5 | 1406 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN359809.1 | - | 346 | Chr. - - [-] on Build |
