siRNA Details
Gene Symbol: LOC100134868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004846.3 | 5 | 1505 | |||
GenBank mRNA | BC030737.1 | - | 5 | 1470 | ||
BC062357.1 | - | 5 | 1470 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN359809.1 | - | 410 | Chr. - - [-] on Build |
