siRNA Details
Gene Symbol: LOC100134868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004846.3 | 5 | 519 | |||
GenBank mRNA | BC030737.1 | - | 5 | 484 | ||
BC062357.1 | - | 5 | 484 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100134868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004846.3 | 5 | 519 | |||
GenBank mRNA | BC030737.1 | - | 5 | 484 | ||
BC062357.1 | - | 5 | 484 |