siRNA Details
Gene Symbol: GHRLOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004431.3 | 5 | 739 | |||
NR_024144.2 | 6 | 567 | ||||
NR_024145.2 | 6 | 911 | ||||
NR_073566.1 | 7 | 922 | ||||
NR_073567.1 | 6 | 910 | ||||
NR_073568.1 | 6 | 765 | ||||
GenBank mRNA | EF549568.1 | - | 4 | 682 | ||
EF549570.1 | - | 4 | 427 | |||
EU072081.1 | - | 3 | 614 | |||
EU072082.1 | - | 2 | 456 | |||
EU789531.1 | - | 3 | 233 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF264800.1 | - | 4 | 648 | Chr. 3 - 10285788 - 10292630 [+] on Build GRCh38 |
