siRNA Details
Gene Symbol: CYMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003599.2 | 2, 3 | 215 | |||
GenBank mRNA | AK310315.1 | - | 3, 4 | 331 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB051751.1 | - | 3, 4 | 331 | Chr. 1 - 110473756 - 110484599 [+] on Build GRCh38 |
