siRNA Details
Gene Symbol: LINC01020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026994.1 | 2 | 222 | |||
GenBank mRNA | BC042977.2 | - | 2 | 224 | ||
BC113532.1 | - | 2 | 118 | |||
BC113534.1 | - | 2 | 118 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX093671.1 | - | 1 | 82 | Chr. 5 - 5038640 - 5038943 [+] on Build GRCh38 |
