siRNA Details
Gene Symbol: TREML3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027256.1 | 1 | 181 | |||
GenBank mRNA | AF534825.1 | - | 1 | 181 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG489188.1 | - | 179 | Chr. - - [-] on Build |
