siRNA Details
Gene Symbol: SPDYE8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003664.2 | 2 | 599 | |||
GenBank mRNA | BC045636.1 | - | 2 | 602 | ||
BC068610.1 | - | 2 | 602 |
Gene Symbol: SPDYE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716210.3 | XP_006716273.1 | 1 | 558 | ||
XM_017012933.1 | XP_016868422.1 | 1 | 562 |
Gene Symbol: SPDYE17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716212.3 | XP_006716275.1 | 1 | 562 | ||
XM_017012888.1 | XP_016868377.1 | 1 | 562 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI562159.1 | - | 2 | 605 | Chr. 7 - 75307380 - 75309220 [+] on Build GRCh38 |
