siRNA Details
Gene Symbol: LINC00652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026883.1 | 4 | 805 | |||
NR_026884.1 | 3 | 628 | ||||
GenBank mRNA | AF161557.1 | - | 5 | 900 | ||
AK094606.1 | - | 3 | 626 | |||
BC029555.1 | - | 3 | 638 |
