siRNA Details
Gene Symbol: ESRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027122.1 | 4 | 2553 | |||
GenBank mRNA | DQ445779.2 | - | 4 | 2552 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ESRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027122.1 | 4 | 2553 | |||
GenBank mRNA | DQ445779.2 | - | 4 | 2552 |