siRNA Details
Gene Symbol: LINC00937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024420.1 | 2 | 510 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI909919.1 | - | 2 | 511 | Chr. 12 - 8356963 - 8390752 [-] on Build GRCh38 |
