siRNA Details
Gene Symbol: PRR26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027151.1 | 1 | 140 | |||
NR_027152.2 | 11 | |||||
GenBank mRNA | AK096000.1 | - | 1 | 140 | ||
AK096013.1 | - | 11 | ||||
AK296168.1 | - | 1 | 55 | |||
BC144473.1 | - | 1 | 111 | |||
DA621677.1 | - | 1 | 140 |
