siRNA Details
Gene Symbol: PRR26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027151.1 | 2 | 350 | |||
NR_027152.2 | 1 | 333 | ||||
GenBank mRNA | AK096000.1 | - | 2 | 350 | ||
AK096013.1 | - | 1 | 333 | |||
AK296168.1 | - | 2 | 265 | |||
BC042561.1 | - | 1 | 144 | |||
BC144473.1 | - | 1 | 433 | |||
DA621677.1 | - | 2 | 350 |
