siRNA Details
Gene Symbol: MIR4697HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024344.1 | 1 | 3977 | |||
GenBank mRNA | AK123849.1 | - | 1 | 2981 | ||
AL133591.1 | - | 1 | 1906 | |||
BC001512.2 | - | 1 | 561 | |||
BC106032.1 | - | 1 | 3838 | |||
BX641055.1 | - | 1 | 3598 |
