siRNA Details
Gene Symbol: LINC01128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047519.1 | 3 | 316 | |||
GenBank mRNA | AK026873.1 | - | 3 | 208 | ||
BC102012.1 | - | 3 | 191 | |||
BC102023.1 | - | 3 | 191 | |||
BM459718.1 | - | 3 | 189 | |||
DR005509.1 | - | 3 | 205 | |||
DR005973.1 | - | 3 | 228 |
