siRNA Details
Gene Symbol: LINC01128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047519.1 | 6 | 1342 | |||
NR_047521.1 | 5 | 1189 | ||||
NR_047523.1 | 4 | 1093 | ||||
NR_047524.1 | 4 | 1005 | ||||
NR_047525.1 | 5 | 1056 | ||||
GenBank mRNA | AK026873.1 | - | 7 | 940 | ||
AK311121.1 | - | 5 | 968 | |||
AK311732.1 | - | 6 | 1043 | |||
BC042880.1 | - | 5 | 1172 |
