siRNA Details
Gene Symbol: AKR1C8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027916.2 | 7 | 758 | |||
GenBank mRNA | AK131335.1 | - | 7 | 760 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AKR1C8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027916.2 | 7 | 758 | |||
GenBank mRNA | AK131335.1 | - | 7 | 760 |