siRNA Details
Gene Symbol: TPTE2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038258.1 | 8 | 4197 | |||
NR_038259.1 | 6 | 4026 | ||||
GenBank mRNA | BC047078.1 | - | 6 | 4026 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TPTE2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038258.1 | 8 | 4197 | |||
NR_038259.1 | 6 | 4026 | ||||
GenBank mRNA | BC047078.1 | - | 6 | 4026 |