siRNA Details
Gene Symbol: TPTE2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038258.1 | 7 | 563 | |||
NR_038259.1 | 5 | 392 | ||||
GenBank mRNA | AK096993.1 | - | 5 | 380 | ||
BC047078.1 | - | 5 | 392 | |||
DA313168.1 | - | 6 | 439 |
