siRNA Details
Gene Symbol: TPTE2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038258.1 | 6, 7 | 531 | |||
NR_038259.1 | 4, 5 | 360 | ||||
GenBank mRNA | AK096993.1 | - | 4, 5 | 348 | ||
BC047078.1 | - | 4, 5 | 360 | |||
DA313168.1 | - | 5, 6 | 407 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA923285.1 | - | 4, 5 | 348 | Chr. 13 - 40800608 - 40921738 [-] on Build GRCh38 |
