siRNA Details
Gene Symbol: TPTE2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038258.1 | 8 | 1546 | |||
NR_038259.1 | 6 | 1375 | ||||
GenBank mRNA | AK093232.1 | - | 3 | 2503 | ||
AK096993.1 | - | 6 | 1363 | |||
BC047078.1 | - | 6 | 1375 |
