siRNA Details
Gene Symbol: WT1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120546.1 | 2 | 1727 | |||
NR_120547.1 | 3 | 1385 | ||||
GenBank mRNA | BC096708.1 | - | 1 | 342 | ||
BC098290.1 | - | 1 | 342 | |||
BC100673.1 | - | 1 | 342 | |||
BX110205.1 | - | 1 | 644 | |||
DQ289489.1 | - | 2 | 1726 | |||
DQ289490.1 | - | 2 | 973 | |||
M60614.1 | - | 1 | 843 |
