siRNA Details
Gene Symbol: WT1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023920.2 | 2 | 957 | |||
NR_120546.1 | 2 | 1427 | ||||
NR_120547.1 | 3 | 1085 | ||||
GenBank mRNA | BC096708.1 | - | 1 | 42 | ||
BC098290.1 | - | 1 | 42 | |||
BC100673.1 | - | 1 | 42 | |||
BX110205.1 | - | 1 | 344 | |||
DQ289488.1 | - | 1 | 2434 | |||
DQ289489.1 | - | 2 | 1426 | |||
DQ289490.1 | - | 2 | 673 | |||
L25110.1 | - | 1 | 4409 | |||
M60614.1 | - | 1 | 543 |
