siRNA Details
Gene Symbol: HAS2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002835.2 | 3 | 1197 | |||
GenBank mRNA | AY941178.1 | - | 3 | 1197 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA336013.1 | - | 2 | 136 | Chr. 8 - 121641436 - 121644693 [+] on Build GRCh38 |
