siRNA Details
Gene Symbol: LINC00051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024378.1 | 3 | 615 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC008253.1 | - | 3 | 615 | Chr. 8 - 142198356 - 142209003 [+] on Build GRCh38 |
