siRNA Details
Gene Symbol: LINC00926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024433.1 | 2 | 1179 | |||
GenBank mRNA | AK090515.1 | - | 1 | 1213 | ||
AK097083.1 | - | 2 | 1179 |
Gene Symbol: PCAT29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097902.1 | - | 2 | 1784 |
