siRNA Details
Gene Symbol: LINC00926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024433.1 | 2 | 1924 | |||
GenBank mRNA | AK097083.1 | - | 2 | 1924 | ||
AK123700.1 | - | 2 | 1300 | |||
AL713736.1 | - | 1 | 383 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024433.1 | 2 | 1924 | |||
GenBank mRNA | AK097083.1 | - | 2 | 1924 | ||
AK123700.1 | - | 2 | 1300 | |||
AL713736.1 | - | 1 | 383 |