siRNA Details
Gene Symbol: EWSAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026949.1 | 3 | 952 | |||
GenBank mRNA | AK091087.1 | - | 3 | 940 | ||
AK097560.1 | - | 4 | 781 | |||
AK292174.1 | - | 3 | 505 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA999185.1 | - | 3 | 505 | Chr. 15 - 69080886 - 69091958 [+] on Build GRCh38 |
