siRNA Details
Gene Symbol: SNORA41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002590.1 | 11 | ||||
GenBank mRNA | AJ609464.1 | - | 11 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA866694.1 | - | 1 | 362 | Chr. 2 - 206161888 - 206162522 [+] on Build GRCh38 |
